EMAIL THIS PAGE TO A FRIEND
Natural product research
Morierinin: a new cytotoxic cucurbitacin from the leaves of Morierina montana Vieill.
PMID 12822899
Abstract
An extract of the leaves of Morierina montana collected in New Caledonia was found to be cytotoxic for KB cells. A bioassay-guided fractionation procedure led to the isolation of a series of cucurbitacins of which one, morierinin (1), is new. The isolation and structure elucidation of 1 are described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0577 Cucurbitacin E, ≥95% (HPLC)
C32H44O8