EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular pharmacology
Effect of cilostazol, a cyclic AMP phosphodiesterase inhibitor, on the proliferation of rat aortic smooth muscle cells in culture.
PMID 1282592
Abstract
Cilostazol, a cyclic AMP phosphodiesterase inhibitor, has been used as an antiplatelet agent. In the present study, we investigated the in vitro effect of cilostazol on DNA synthesis in rat aortic arterial smooth muscle cells (SMCs) in culture stimulated with fetal calf serum (FCS), platelet-derived growth factor (PDGF), insulin, or insulin-like growth factor-I (IGF-I). Micromolar concentrations of cilostazol inhibited [3H]thymidine incorporation into DNA and cell growth as determined by cell number and protein concentration. Treatment with cilostazol increased the intracellular concentration of cyclic AMP, suggesting that the inhibition of SMC proliferation by cilostazol may be mediated through increased levels of cyclic AMP. The results suggested that cilostazol, by interfering with the proliferation of arterial SMCs, may have potential to prevent initiation and progression of atherosclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1134153
Cilostazol, United States Pharmacopeia (USP) Reference Standard
C20H27N5O2