EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Effect of olvanil on the afferent and efferent function of capsaicin-sensitive C-fibers in guinea pig airways.
PMID 12826239
Abstract
The aim of the present study was to examine the ability of the nonpungent vanilloid VR1 receptor agonist, olvanil, to activate the afferent and efferent function of capsaicin-sensitive C-fibers in guinea pig airways. We found that while capsaicin (10 nM-10 microM) and resiniferatoxin (0.1 nM-1.0 microM) evoked a robust contraction of the guinea pig trachea in vitro, olvanil (10 nM-10 microM) was a weak spasmogen. In addition, pretreatment with olvanil caused only a minor reduction of subsequent responses to capsaicin or resiniferatoxin. Using single fiber recording from guinea pig airway C-fibers, we found that olvanil (10 microM) did not evoke action potential discharge although these fibers responded vigorously to capsaicin after prolonged treatment with olvanil (10 microM). These findings are indicative of significant differences in the relative sensitivity of vanilloid VR1 receptor-transfected cells and the peripheral terminals of airway C-fibers to pungent and nonpungent vanilloid VR1 receptor agonists.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0257
Olvanil, powder
C26H43NO3