EMAIL THIS PAGE TO A FRIEND
Plant & cell physiology
Redox changes in the chloroplast and hydrogen peroxide are essential for regulation of C(3)-CAM transition and photooxidative stress responses in the facultative CAM plant Mesembryanthemum crystallinum L.
PMID 12826622
Abstract
Mesembryanthemum crystallinum, a facultative halophyte and C(3)-Crassulacean acid metabolism (CAM) intermediate plant, has become a favoured plant for studying stress response mechanisms during C(3)-CAM shifts. One hour of exposure to excess light (EL) caused inhibition of photosynthetic electron transport in M. crystallinum leaves as indicated by chlorophyll a fluorescence measurements. This was accompanied by an increase in NADP-malic enzyme (ME), one of the key cytosolic enzymes involved in CAM, and by a general increase in superoxide dismutase (SOD) activity. In contrast, NAD-ME activity (the mitochondrial form of ME) was not affected by EL. Exposure to EL and 2,5-dibromo-3-methyl-6-isopropyl-p-benzoquinone (DBMIB) treatment of a whole plant in low light induced hydrogen peroxide (H(2)O(2)) and C(3) to CAM transition. In contrast, treatment with 3-3,4-dichlorophenyl-1,1-dimethyl urea (DCMU) has blocked high light-induced H(2)O(2) accumulation and C(3)-CAM transition. Moreover, the abundance of transcripts encoding different SODs, ascorbate peroxidase and SOD activity was differently regulated by DCMU and DBMIB. Results of applying EL or high light, H(2)O(2) and photosynthetic electron transport inhibitors suggest that the redox events in the vicinity of PSII and/or PSI and photo-produced H(2)O(2) play a major role in the regulation of C(3)-CAM transition and photooxidative stress responses in M. crystallinum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
271993
2,5-Dibromo-6-isopropyl-3-methyl-1,4-benzoquinone
C10H10Br2O2