EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Sensitive determination of pethidine in body fluids by gas chromatography-tandem mass spectrometry.
PMID 12829004
Abstract
We have presented a simple and sensitive method for determining pethidine, a narcotic analgesic drug in body fluids by gas chromatography-tandem mass spectrometry (GC-MS/MS). Pethidine and 4'-piperidinoacetophenone (internal standard) were extracted from body fluids with Bond Elut C(18) columns; the recoveries were above 85% for both compounds. The calibration curves for blood and urine showed good linearities in the range of 1.25-40 ng/ml. Its detection limits (signal-to-noise ratio=3) were estimated to be approximately 0.5 ng/ml of whole blood and urine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119725
4′-Piperidinoacetophenone, 97%
C13H17NO