EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
Requirement of Watson-Crick hydrogen bonding for DNA synthesis by yeast DNA polymerase eta.
PMID 12832493
Abstract
Classical high-fidelity DNA polymerases discriminate between the correct and incorrect nucleotides by using geometric constraints imposed by the tight fit of the active site with the incipient base pair. Consequently, Watson-Crick (W-C) hydrogen bonding between the bases is not required for the efficiency and accuracy of DNA synthesis by these polymerases. DNA polymerase eta (Poleta) is a low-fidelity enzyme able to replicate through DNA lesions. Using difluorotoluene, a nonpolar isosteric analog of thymine unable to form W-C hydrogen bonds with adenine, we found that the efficiency and accuracy of nucleotide incorporation by Poleta are severely impaired. From these observations, we suggest that W-C hydrogen bonding is required for DNA synthesis by Poleta; in this regard, Poleta differs strikingly from classical high-fidelity DNA polymerases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
133892
2,4-Difluorotoluene, 99%
C7H6F2