EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Spectrofluorometric determination of trace amounts of terbium with 4-chlorosalicylic acid, EDTA, and cetyltrimethylammonium bromide.
PMID 12834236
Abstract
The quadruple complex formed by terbium with 4-chlorosalicylic acid (CSA), EDTA and cetyltrimethylammonium bromide (CTMAB) has been used for the sensitive spectrofluorometric determination of terbium in mixed rare earths. The effect of the experimental conditions on the fluorescence intensity was defined. Under the optimum conditions selected, the fluorescence intensity was linear with the terbium concentration in the range of 3.0 x 10(-8)-1.0 x 10(-5) mol/L with a detection limit of 8.0 x 10(-9) mol/L (S/N = 3). It has been satisfactory for the determination of terbium in mixed rare earths with good recovery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183784
4-Chlorosalicylic acid, 93%
C7H5ClO3