EMAIL THIS PAGE TO A FRIEND
Biological trace element research
Zirconium. An abnormal trace element in biology.
PMID 1283692
Abstract
The action of Zirconium (Zr) on biological systems presents an enigma. It is ubiquitous, being present in nature in amounts higher than most trace elements. It is taken up by plants from soil and water and accumulated in certain tissues. The entry into animal systems in vivo is related to the mode of exposure and the concentration in the surrounding environment. Retention is initially in soft tissues and then slowly in the bone. The metal is able to cross the blood brain-barrier and is deposited in the brain and the placental barrier to enter milk. The daily human uptake has been known to be as high as 125 mg. The level of toxicity has been found to be moderately low, both in histological and cytological studies. The toxic effects induced by very high concentrations are nonspecific in nature. Despite the presence and retention in relatively high quantities in biological systems, Zr has not yet been associated with any specific metabolic function. Very little information is available about its interaction with the compounds of the genetical systems, such as nucleic acids. Apparently, the metal is neither an essential nor toxic element in the conventional sense. However, the increasing exposure to this element through its increasing use in new materials and following radioactive fallout, has increased the importance of the study of its effects on living organisms. The tetravalent nature of the ionic state and the high stability of the compounds formed are important factors that need to be considered, as also the accumulation of this element in the brain, reminiscent of the relationship between Al3+ and Alzheimer's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF84779612
Zirconium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%
Zr
GF53330172
Zirconium, tube, 1000mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%
Zr
GF56980165
Zirconium, rod, 1000mm, diameter 12.7mm, centerless ground, 99.2%
Zr
GF03685316
Zirconium, foil, 100x100mm, thickness 0.1mm, annealed, 99.2%
Zr
GF86559384
Zirconium, foil, 1m coil, thickness 0.05mm, annealed, 99.2%
Zr
GF69451487
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zr
GF50488577
Zirconium, foil, 100x100mm, thickness 0.5mm, annealed, 99.2%
Zr
GF11484468
Zirconium, rod, 1000mm, diameter 9.5mm, centerless ground, 99.2%
Zr
GF36220227
Zirconium, rod, 1000mm, diameter 5.0mm, centerless ground, 99.2%
Zr
GF50035137
Zirconium, foil, 5m coil, thickness 0.025mm, annealed, 99.2%
Zr
GF83603909
Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%
Zr
GF04513700
Zirconium, rod, 100mm, diameter 50mm, centerless ground, 99.2%
Zr
GF10742284
Zirconium, foil, 150x150mm, thickness 2.0mm, annealed, 99.2%
Zr
GF74069837
Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%
Zr
GF29806982
Zirconium, foil, light tested, 200x200mm, thickness 0.01mm, 99.2%
Zr
GF63904989
Zirconium, foil, light tested, 100x100mm, thickness 0.025mm, annealed, 99.2%
Zr
GF84948834
Zirconium, foil, 0.5m coil, thickness 0.075mm, annealed, 99.2%
Zr
GF04668835
Zirconium, foil, 25x25mm, thickness 0.5mm, annealed, 99.2%
Zr
GF30135002
Zirconium, sponge, 100g, max. size 25mm, 99.2%
Zr
GF88851730
Zirconium, tube, 100mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%
Zr
GF45997925
Zirconium, foil, light tested, 25x25mm, thickness 0.006mm, 99.2%
Zr
GF27595637
Zirconium, rod, 100mm, diameter 6.0mm, centerless ground, 99.2%
Zr
GF90874121
Zirconium, foil, 6mm disks, thickness 0.1mm, annealed, 99.2%
Zr
GF43362660
Zirconium, foil, 1m coil, thickness 0.02mm, 99.2%
Zr
GF26818971
Zirconium, wire reel, 1m, diameter 0.5mm, as drawn, 99.2%
Zr
GF03886443
Zirconium, foil, 10mm disks, thickness 0.5mm, annealed, 99.2%
Zr
GF99338743
Zirconium, rod, 200mm, diameter 2.0mm, centerless ground, 99.2%
Zr
GF87053474
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zr
GF43414556
Zirconium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.2%
Zr
GF66876014
Zirconium, sponge, 200g, max. size 25mm, 99.2%
Zr