EMAIL THIS PAGE TO A FRIEND
Journal of the American College of Cardiology
Thromboxane inhibition improves renal perfusion and excretory function in severe congestive heart failure.
PMID 12849673
Abstract
The aim of this study was to evaluate whether thromboxane inhibition can favorably affect renal perfusion and clinical conditions in patients affected by severe heart failure. The renal formation of the vasoconstrictor thromboxane A(2) (TxA(2)) is increased during cardiac failure. By oral administration of picotamide (a renal TxA(2) synthase and TxA(2)/prostaglandin H(2) receptor inhibitor), we blocked renal TxA(2). Fourteen patients in New York Heart Association functional class IV were studied according to a randomized, double-blinded, cross-over design. Each of the two eight-day periods of testing was preceded by a three-day period during which certain vasoactive medications were stopped. Daily 24-h total urinary thromboxane B(2) (TxB(2)), the stable metabolite of TxA(2), dropped at the end of picotamide treatment (p < 0.01 vs. baseline). Compared with placebo, effective renal plasma flow and the glomerular filtration rate increased (p < 0.01 and p < 0.05, respectively), thus leading to a significant decrease in the filtration fraction (p < 0.01). Renal vascular resistance decreased consistently (p < 0.01). In all patients, picotamide treatment was associated with an increase in diuresis and natriuresis (p < 0.001 vs. baseline). Plasma creatinine decreased (p < 0.05 vs. baseline). Patients also showed improvement in several clinical parameters, including a significant decrease in both pulmonary and venous pressure (p < 0.01 vs. baseline). These results indicate that renal thromboxane formation plays an important role in renal vascular resistance in patients with severe heart failure, such as those described in the present study. Inhibition of TxA(2) improves renal hemodynamics and kidney function and favorably affects indexes of cardiac performance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8477
Picotamide
C21H20N4O3