EMAIL THIS PAGE TO A FRIEND
Parasitology research
In vitro trypanocidal activity of dibutyltin dichloride and its fatty acid derivatives.
PMID 12851812
Abstract
Searching for new compounds against pathogenic trypanosomes has been substantially accelerated by the development of in vitro screening assays. In an attempt to explore the chemotherapeutic potential of organotin compounds and to broaden the search for newer trypanocides, fatty acid derivatives of dibutyltin dichloride were synthesized and their in vitro trypanocidal profiles studied on Trypanosoma brucei brucei, Trypanosoma brucei gambiense and Trypanosoma brucei rhodesiense. A 24-h time course experiment was conducted with various concentrations of the compounds using a 24-well microtiter plate technique. The compounds tested were trypanocidal in a dose-dependent fashion: inhibiting survival and growth, resulting in irreversible morphological deformation and the eventual death of the parasites. The minimum inhibitory concentrations of the tested diorganotins are at low micromolar ranges: from 0.15-0.75 microM for T. b. brucei, T. b. gambiense and T. b. rhodesiense. These observations suggest that organotin has chemotherapeutic potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
291234
Dibutyltin dilaurate, 95%
C32H64O4Sn