EMAIL THIS PAGE TO A FRIEND
Nature
The DNA sequence of human chromosome 7.
PMID 12853948
Abstract
Human chromosome 7 has historically received prominent attention in the human genetics community, primarily related to the search for the cystic fibrosis gene and the frequent cytogenetic changes associated with various forms of cancer. Here we present more than 153 million base pairs representing 99.4% of the euchromatic sequence of chromosome 7, the first metacentric chromosome completed so far. The sequence has excellent concordance with previously established physical and genetic maps, and it exhibits an unusual amount of segmentally duplicated sequence (8.2%), with marked differences between the two arms. Our initial analyses have identified 1,150 protein-coding genes, 605 of which have been confirmed by complementary DNA sequences, and an additional 941 pseudogenes. Of genes confirmed by transcript sequences, some are polymorphic for mutations that disrupt the reading frame.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K1502 α-Ketoglutarate Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 0.1-1.0 units/mg protein (Lowry)
C2888
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
C0663
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)
AV54470
Anti-SUNC1 antibody produced in rabbit, affinity isolated antibody
D2197 Diaphorase from Clostridium kluyveri, recombinant, expressed in E. coli, Animal-component free
D5540 Diaphorase from Clostridium kluyveri, lyophilized powder, 3.0-20.0 units/mg protein (biuret)
D2322 Diaphorase from Clostridium kluyveri, recombinant, expressed in E. coli, Animal-component free
I1782 Inosine Monophosphate Dehydrogenase Type II human, recombinant, expressed in E. coli
M9004 Malic Dehydrogenase from bovine heart, ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)
M2634 Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
M7383 Malic Dehydrogenase from porcine heart, ≥400 units/mg protein (biuret), ammonium sulfate suspension
M1567 Malic Dehydrogenase from porcine heart, ≥600 units/mg protein (biuret), ammonium sulfate suspension
A1705 S-(5′-Adenosyl)-L-homocysteine Hydrolase from rabbit erythrocytes, buffered aqueous glycerol solution, ≥10 units/mg protein (Lowry)