EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on chemical constituents of Choerospondias axillaris].
PMID 12858769
Abstract
Ten compounds were isolated from the dry fruit of Choerospondias axillaris (Roxb.) Burtt et Hill. Their structures were elucidated as dihydroquercetin(1), quercetin(2), protocatechuic acid(3), gallic acid(4), 3,3'-di-o-methylellagic acid(5), beta-sitosterol(6), daucosterol (7), stearic acid(8), triacontanoic acid(9), octacosanol(10) by IR, EI-MS and NMR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3379
1-Octacosanol, ≥99% (GC)
C28H58O