EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Success of prostaglandin E2 in structure-function is a challenge for structure-based therapeutics.
PMID 12861081