EMAIL THIS PAGE TO A FRIEND
Blood cells
Review: the platelet storage lesion: possible role of plasticizers?
PMID 1286197
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
538140
Trioctyl trimellitate, 99%
C33H54O6
92124
Tris(2-ethylhexyl) trimellitate, Selectophore
C33H54O6