EMAIL THIS PAGE TO A FRIEND
Organic letters
Efficient, stereoselective synthesis of oxazolo[3,2-a]pyrazin-5-ones: novel bicyclic lactam scaffolds from the bicyclocondensation of 3-aza-1,5-ketoacids and amino alcohols.
PMID 12868900
Abstract
[reaction: see text] The bicyclocondensation of 3-aza-1,5-ketoacids and amino alcohols furnished novel oxazolo[3,2-a]pyrazin-5-one scaffolds possessing angular, ring junction substituents in high yield with excellent levels of substrate-based diastereocontrol. Mild oxidation of serinol-derived scaffolds provided access to a new class of constrained dipeptide surrogates. Deprotection of the endocyclic amine contained within these scaffolds allows for further diversification via N-functionalization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357898
Serinol, 98%
C3H9NO2