EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Macromolecular specificity of collagen fibrillogenesis: fibrils of collagens I and XI contain a heterotypic alloyed core and a collagen I sheath.
PMID 12869566