EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Mass spectrometric studies of cyclopentanol derivatives in the reductive coupling of alpha,beta-unsaturated ketones assisted by samarium diiodide.
PMID 12872273
Abstract
The mass spectrometric fragmentations of a series of cyclopentanol derivatives in positive fast-atom bombardment (FAB(+)) mode were investigated. The corresponding pathways proposed were confirmed by MS/MS data obtained using linked scans at constant B/E, high-resolution accurate mass data, and comparisons with corresponding information for a specifically deuterated molecule.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C112208
Cyclopentanol, 99%
C5H10O