EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Quinaldic acid as a new matrix for matrix-assisted laser desorption/ionization of nucleic acids.
PMID 12872286
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160660
Quinaldic acid, 98%
C10H7NO2