EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Facile preparation of DNA-tagged carbohydrates.
PMID 12873482
Abstract
We report here that unprotected carbohydrates (maltose, lactose, cellobiose, and maltoheptaose) can be attached to the aminoalkylated oligonucleotides under mild reductive-amination conditions (aqueous borate buffer, pH 8.0, NaBH(3)CN, 60 degrees C) without notable side reactions. Quadruplex-forming G-rich oligonucleotide, 5'-aminoalkyl d(TGGGGT), is glycosylated with maltoheptaose to afford a novel DNA-assisted tetrasaccharide cluster motif.