EMAIL THIS PAGE TO A FRIEND
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie
Glucuronidation of 4-methylumbelliferone and 4-hydroxybiphenyl and in vitro induction of UDP-glucuronosyltransferase 2B12-mRNA in precision-cut rat liver slices.
PMID 12877362
Abstract
Fresh rat liver slices were used to demonstrate the glucuronidation of the model substrates 4-methylumbelliferone (MU) and 4-hydroxybiphenyl (HB). Both glucuronidation reactions proved to be more stable than cytochrome P450-dependent monooxygenations. After an incubation time of 48 h there was no decrease in MU glucuronidation rate, whereas HB glucuronidation was stable until 24 h, and then decreased by about 50% until 48 h. The technique of quantitative competitive RT-PCR was used to determine the expression of UDP-glucuronosyltransferase 2B12-mRNA (UGT2B12-mRNA) in precision-cut rat liver slices. Constitutive levels of UGT2B12-mRNA were measurable. Following 24 h culture of rat liver slices in the presence of phenobarbital, the level of UGT2B12-mRNA increased about twofold, which corresponds to the inducibility in vivo. The addition of beta-naphthoflavone had no influence. The results show that precision-cut liver slices are not only suitable for the detection of an in vitro induction of cytochrome P450-mRNAs, which is characterized by high induction factors, but also of poor induction effects, e.g. on UGT2B12-mRNA, provided that the respective mRNA is exactly quantified.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
424625
4-Phenylphenol, purified by sublimation, 99%
C12H10O
134341
4-Phenylphenol, 97%
C12H10O