EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Bis-functionalized fullerene-dibenzo[18]crown-6 conjugate: synthesis and cation-complexation dependent redox behavior.
PMID 12877537
Abstract
A one-step synthesis of bis-pyrrolidine functionalized fullerene-dibenzo[18]crown-6 conjugate and its metal cation complexation to the crown ether entity dependent redox behavior is reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158399
Dibenzo-18-crown-6, 98%
C20H24O6
49902
Tin(II) ionophore I, Selectophore
C20H24O6