EMAIL THIS PAGE TO A FRIEND
Food additives and contaminants
Lack of de-epoxidation of type B trichothecenes in incubates with human faeces.
PMID 12881132
Abstract
The ability of human gastrointestinal organisms to transform the trichothecenes 3-acetyldeoxynivalenol and nivalenol was investigated. Samples of human faeces were incubated under anaerobic conditions for 48 h with the toxins. They were then extracted and analysed for trichothecenes and metabolites. 3-acetyldeoxynivalenol was metabolized to deoxynivalenol during the incubation period. In contrast to what has been reported for other species such as rats, mice and pigs, no de-epoxidated metabolites were detected in the faecal incubates. The toxicological significance of the difference in the intestinal ability to transform trichothecenes between species is unknown.