EMAIL THIS PAGE TO A FRIEND
Roczniki Panstwowego Zakladu Higieny
[Comparison of auc values for caffeine and its metabolites after single and repeated administration of 2-methoxy-5-nitrobenzyl bromide].
PMID 12884508