EMAIL THIS PAGE TO A FRIEND
Obstetrical & Gynecological Survey
Performance of visual inspection with acetic acid for cervical cancer screening: a qualitative summary of evidence to date.
PMID 12886165