EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series
Introduction of fluorine into the sugar moiety of nucleoside using diethylaminosulfur trifluoride (DAST).
PMID 1289833
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235253
(Diethylamino)sulfur trifluoride, 95%
C4H10F3NS