EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Derivatization reaction of the mycotoxin moniliformin with 1,2-diamino-4,5-dichlorobenzene: structure elucidation of an unexpected reaction product by liquid chromatography/tandem mass spectrometry and liquid chromatography/nuclear magnetic resonance spectroscopy.
PMID 12898650
Abstract
The derivatization reaction of the mycotoxin moniliformin with 1,2-diamino-4,5-dichlorobenzene was previously introduced to improve distinctly the sensitivity of an assay applying high-performance liquid chromatography prior to fluorescence detection. In the course of the implementation of this derivatization approach into a liquid chromatographic/mass spectrometric method, an unexpected derivatization product has now been discovered by mass spectrometry. In order to elucidate its structure, detailed investigations with liquid chromatography/tandem mass spectrometry and liquid chromatography coupled on-line with NMR spectroscopy were performed. These studies give evidence for a heterocyclic structure that has been formed by the loss of one water and one carbon monoxide molecule. A reasonable mechanism for this derivatization reaction is proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D71607
4,5-Dichloro-o-phenylenediamine, 97%
C6H6Cl2N2