EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Use of chiral zwitterionic surfactants for enantiomeric resolutions by capillary electrophoresis.
PMID 12900862
Abstract
The enantiomeric resolution of 1,1'-binaphthyl-2,2'-diamine and Tröger's base was investigated using the commercially available zwitterionic surfactants 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulphonate (CHAPS) and 3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulphonate (CHAPSO). Resolution of the weakly basic chiral probes was achieved using varying concentrations of surfactant, above their critical micellar concentrations, in a phosphate buffer (pH 2.5; 100 mM) to ensure ionisation of the analytes. Both CHAPS and CHAPSO were employed in the absence of additional coselectors or surfactants as sole micellar-forming agents. The addition of organic modifiers, methanol and acetonitrile (ACN), to the background electrolyte (BGE) was found to have a detrimental effect on enantioselectivity presumably by alteration of the phase polarity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3649
CHAPSO, ≥98%
C32H58N2O8S
C4695
CHAPSO, BioXtra
C32H58N2O8S