EMAIL THIS PAGE TO A FRIEND
Pharmaceutical development and technology
Relationship between internal phase volume and emulsion stability: the cetyl alcohol/stearyl alcohol system.
PMID 12901692
Abstract
The main objective of this study was to optimize the stability of cetyl alcohol/stearyl alcohol emulsions in terms of percentage of internal phase volume, emulsifier type and concentration, and amount of external phase (water). Creams (o/w emulsions) were prepared by phase inversion and physical properties as particle size of the internal phase, apparent viscosity, and sedimentation volume evaluated. Stability was performed at room temperature, 40 degrees C, 50 degrees C, and under stress conditions. High hydrophilic lipophilic balance (HLB) nonionic surfactants as tween 80, tween 20, Myrj 52, Brij 35, and low HLB span 60 were used as emulsifying agents. The percentage of internal phase components (cetyl alcohol and stearyl alcohol), percentage of emulsifying agents, and percentage of aqueous external phase were varied, and stability was investigated. As the level of emulsifier agent (tween 80 and span 60) increased from 3% to 15%, and the percent of the internal phase remained constant at 30%, the particle size of the internal phase decreased and the cream became more stable. Formulations of the same composition, but prepared using Myrj 53 and tween 20 as emulsifiers, showed a larger particle size than formulations prepared using tween 80 and Brij 35. As the level of the internal phase volume increased and consequently the amount of water decreased, emulsion viscosity increased. The best formulation containing 30% internal phase (50% cetyl alcohol, 35% stearyl alcohol), 15% emulsifying agents (tween 80/span 60 ratio of 3:1), and 70% water was selected, and effects of process temperature and cooling rate on emulsion stability investigated. This formulation was further investigated in terms of stability of a 1% hydrocortisone addition by varying the percentage (30%, 35%, 40%, and 45%) of internal phase and percentage of water (70%, 65%, 60%, and 55%). The best formulation contained 45% internal phase (22.5 g cetyl alcohol, 15.75 g stearyl alcohol, 15% emulsifying agent, which is equivalent to 5 g tween 80 and 1.7 g span 60), and 55% w/w water, was manufactured under different manufacturing processes. Emulsions prepared by homogenization at the beginning of the process of emulsification were stable with small internal phase particle diameter. This study demonstrates that at every cetyl alcohol/stearyl alcohol ratio there is a phase volume/emulsifier HLB ratio, which results in optimum stability.