EMAIL THIS PAGE TO A FRIEND
Archives of histology and cytology
Preparation of human chromosomes for high resolution scanning electron microscopy.
PMID 1290676
Abstract
The addition of ethidium bromide during the last 2.5-3 h of lymphocyte culturing restricted chromosome contraction and preserved the banding structure in scanning electron microscopy. Treatment of the chromosomes with trypsin and use of impregnation with osmium tetroxide and thiocarbohydrazide resulted in a structural preservation of high resolution quality.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223220
Thiocarbohydrazide, 98%
CH6N4S