EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Preparation of graphitic carbon nanofibers with the use of water-soluble supports.
PMID 12908314
Abstract
Graphitic carbon nanofibers (GCNFs) are prepared from CO/H2 with a nickel-rich Fe-Ni (2:8) growth catalyst supported on three different water-soluble supports (Na4SiO4, Na2SiO3, and Na2CO3). GCNF products are formed with yield and crystallinity comparable to those of GCNF product produced with the same growth catalyst supported on fumed silica. Separation of GCNF product from the solid support is accomplished by aqueous dissolution of the support. This synthetic method extends the convenient isolation of GCNFs formed with supported growth catalysts to a wide variety of potential solid supports.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
307815
Sodium metasilicate
Na2O3Si