EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Determination of taurine and hypotaurine in animal tissues by reversed-phase high-performance liquid chromatography after derivatization with dabsyl chloride.
PMID 12908604
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
39068
4-(Dimethylamino)azobenzene-4′-sulfonyl chloride, ≥97.5% (AT)
C14H14ClN3O2S