EMAIL THIS PAGE TO A FRIEND
Applied occupational and environmental hygiene
Folpet permeation through nitrile gloves.
PMID 12909534
Abstract
The aim of this study was to investigate whether two different brands of unsupported and unlined nitrile gloves protected against aqueous emulsions of a Folpet wettable powder (50% Folpet) using an ASTM type-I-PTC 600 permeation cell at 30.0 +/- 0.1 degrees C held in a shaking water bath. An analytical method to determine Folpet using the internal standard method was first developed based on gas chromatography-mass spectrometry (GC-MS), and gas chromatography-electron capture detection (GC-ECD). A novel pyrolysis GC-ECD technique that quantified the thermal degradation product phthalimide had pg sensitivity suitable to detect the trace amounts of Folpet that permeated. The on-column conversion was (68.0 +/- 9.5) percent at 170 degrees C over the folpet injected mass range of 3 to 148 pg. The challenge solution in the permeation cell was 1.4 mg/mL aqueous emulsion of Folpet wettable powder, and 2-propanol was the collection solvent. After evaporation of the collection solvent, the time weighted average rate of permeation of Folpet through SafeSkin nitrile (an exams type of glove) after 8 hours was (42.1 +/- 2.9) ng/cm(2)/min compared with (2.04 +/- 0.69) ng/cm(2)/min for the Sol-Vex nitrile (industrial chemical resistant), the latter being about 21 times more protective and also near the limits of detection. The respective values after 4 hours of exposure were (28.4 +/- 1.2) and (0.65 +/- 0.36) ng/cm(2)/min. Diagnostic reflectance infrared minima of both challenge and collection sides of the gloves showed small changes in wave number and intensity values after 8 hours of exposure, with Folpet being detected in dried spots on the challenge side. GC-ECD-based permeation and IR reflectance data indicated high chemical resistance of the Sol-Vex gloves to an aqueous emulsion of Folpet.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32057
Folpet, PESTANAL®, analytical standard
C9H4Cl3NO2S