EMAIL THIS PAGE TO A FRIEND
Guang pu xue yu guang pu fen xi = Guang pu
[Photolysis kinetics and photoinitiated polymerization mechanism studies of [Cp-Fe-Naph]BF4 and [Cp-Fe-Cp]BF4 by UV spectrum].
PMID 12914164
Abstract
The absorption spectra of photoinitiator cyclopentadiene-Fe-naphthalene tetrafluoroborate salt ([Cp-Fe-Naph]BF4) and ferocinium tetrafluoroborate salt([Cp-Fe-Cp]BF4), in two solvents have been studied. The influence of irradiation on the solution of photoinitiator has been observed. The kinetics of photolysis has been determined by real-time UV absorption spectrum. The results show that the photolysis reactions of two initiators are the first order reaction, the rate constant of photolysis reaction are 0.72 and 0.65 min-1, respectively. The photoinitiated mechanism of [Cp-Fe-Cp]BF4 was obtained.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388297
Ferrocenium hexafluorophosphate, 97%
C10H10F6FeP
482358
Ferrocenium tetrafluoroborate, technical grade
C10H10BF4Fe