EMAIL THIS PAGE TO A FRIEND
Journal of chemical ecology
Identification of the female-produced sex pheromone of the scarab beetle, Hoplia equina.
PMID 12921441
Abstract
Hoplia equina LeConte (Coleoptera Scarabaeidae: Melolonthinae) is a beetle pest of cranberry beds in Massachusetts. Larvae feed on the roots of the cranberry plant, reducing yield as well as vine density. The female sex pheromone was identified as 2-tetradecanone. There were eight compounds found in the airborne volatiles collected from females that elicited antennal responses from males. Of the eight compounds tested (nonanal, decanal, dodecanal, 2-dodecanone, 2-tridecanone, 2-tetradecanone, 2-pentadecanone, and 2-hexadecanone), 2-tetradecanone was the only one that attracted male beetles in the field. Combining any of the other seven antennally active compounds with 2-tetradecanone did not increase male capture.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
374180
2-Hexadecanone, 98%
C16H32O