EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of the single component and competitive adsorption isotherms of the 1-indanol enantiomers by the inverse method.
PMID 12924781
Abstract
The inverse method of isotherm determination consists in calculating the numerical values of the coefficients of an isotherm model that give a set of chromatographic profiles in best possible agreement with the set of experimental profiles available. This method was applied to determine the adsorption isotherms of the 1-indanol enantiomers on a cellulose tribenzoate chiral stationary phase. Both single-component and competitive isotherms were determined by using no more than one or two overloaded band profiles. The isotherms determined from the overloaded band profiles agreed extremely well with the isotherms determined by frontal analysis. Several isotherm models were used and tested. The best-fit isotherm was selected by means of statistical evaluation of the results. The results show that the adsorption is best characterized with a model describing heterogeneous adsorption with bimodal adsorption energy distribution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328413
(R)-(−)-1-Indanol, 99%
C9H10O
323128
(S)-(+)-1-Indanol, 99%
C9H10O
193739
1-Indanol, 98%
C9H10O