EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of (R)-2-methyl-4-deoxy and (R)-2-methyl-4,5-dideoxy analogues of 6-phosphogluconate as potential inhibitors of 6-phosphogluconate dehydrogenase.
PMID 12926259
Abstract
The synthesis of (2R)-2-methyl-4,5-dideoxy and (2R)-2-methyl-4-deoxy analogues of 6-phosphogluconate is described. The synthetic strategy relies on the Evans aldol reaction for the installation of the chiral centres in the 2- and 3-positions. The selective phosphorylation at the primary alcohol function of (2R,3S)-3,6-dihydroxy-2-methylhexanoic acid benzyl ester (5) and (2R,3S,5S)-3,5,6-trihydroxy-2-methylhexanoic acid benzyl ester (20) was achieved with dibenzyl phosphochloridate and dibenzyl phosphoiodinate respectively, working at low temperature. (2R,3S)-3-Hydroxy-2-methyl-6-phosphonoxyhexanoic acid (9) was obtained in 25% overall yield from 4-benzyloxybutanol and (2R,3S,5S)-3,5-dihydroxy-2-methyl-6-phosphonoxyhexanoic acid (28) in 10% overall yield from L-malic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7627
6-Phosphogluconic acid barium salt hydrate, Grade VI, ≥90%, powder
C6H10Ba1.5O10P · xH2O