EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Adsorption and desorption of triasulfuron by soil.
PMID 12926882
Abstract
The adsorption and desorption of the herbicide triasulfuron [2-(2-chloroethoxy)-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide] by three soils, soil organic matter (H(+) and Ca(2+)-saturated), and an amorphous iron oxide were studied. Adsorption isotherms conformed to the Freundlich equation. It was found that pH is the main factor influencing the adsorption in all of the systems. Indeed, the adsorption on soils was negatively correlated with pH. The highest level of adsorption was measured on soils with low pH and high organic carbon content. Moreover, it was found that humic acid is more effective in the adsorption compared with calcium humate (the pH values of the suspensions being 3.5 and 6, respectively). Experiments on amorphous iron oxide confirmed the pH dependence. Desorption was hysteretic on soils having high organic carbon content.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33383
Triasulfuron, PESTANAL®, analytical standard
C14H16ClN5O5S