EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Extraction and formation dynamic of oak-related volatile compounds from different volume barrels to wine and their behavior during bottle storage.
PMID 12926895
Abstract
The extraction rate of furfuryl aldehydes, guaiacol, and 4-methylguaiacol, cis- and trans-oak lactone, and vanillin and the formation rate of furfuryl alcohol and the volatile phenols 4-ethylguaiacol and 4-ethylphenol have been studied in wines matured in different capacity oak barrels (220, 500, and 1000 L). Also, the behavior of these compounds during 1 year of wine bottle storage was followed. The lactones were extracted at a linear rate with large differences that depended on barrel volume. Those compounds related to oak toasting (guaiacol, 4-methylguaiacol, furfuryl aldehydes, and vanillin) seemed to be extracted faster during the first days of oak maturation except for vanillin, which required at least 3 months to accumulate in the wine. The volatile phenols, 4-ethylphenol and 4-ethylguaiacol, were formed in large quantities after the first 90 days of oak maturation, coinciding with the end of spring and beginning of summer. Wines matured in 1000-L oak barrels resulted in the lowest levels of volatile compound accumulation. During bottle storage, some compounds decreased in their concentration (5-methylfurfural, vanillin), others experienced increases in their levels (lactones, furfural, 4-ethylguaiacol, 4-ethylphenol), and the concentration of other compounds hardly changed (guaiacol, furfuryl alcohol).