EMAIL THIS PAGE TO A FRIEND
Toxicology
Allergenicity evaluation of p-chloro-m-cresol and p-chloro-m-xylenol by non-radioactive murine local lymph-node assay and multiple-dose guinea pig maximization test.
PMID 12927380
Abstract
p-Chloro-m-cresol (PCMC) and p-chloro-m-xylenol (PCMX) are known to cause allergic contact dermatitis. For risk assessment of skin sensitizers, information on dose-response profiles in the induction and elicitation phases and cross-reactivity with analogous chemicals are important. In the non-radioactive local lymph-node assay (LLNA) using 5-bromo-2'-deoxyuridine instead of 3H-methyl thymidine, significant effect on lymph node cell proliferation was detected at 10% PCMC and 25% PCMX, while in the multiple-dose guinea pig maximization test (GPMT) at least one animal tested in the group was sensitized at a 5 ppm induction dose of either chemical. When mean skin reaction score in an animal group maximally sensitized with each allergen with the GPMT was plotted against log challenge concentration, linear regression lines with high correlations were obtained in both cases. The calculated elicitation threshold was lower for PCMC than PCMX. The area under the linear regression line between the threshold point and 1% of the elicitation concentration, another index of relative elicitation potency, was also greater for PCMC. Bidirectional cross-reactivity between PCMX and PCMC was detected in the GPMT. PCMC was thus identified in both LLNA and GPMT as a stronger sensitizer than PCMX in both the induction and elicitation phases. These results suggest that the non-radioactive LLNA is a simple and useful method for evaluating allergenicity in the induction phase, while the GPMT using a maximally sensitized animal group is more suitable for assessing the dose-response profile and cross-reactivity in the elicitation phase.
Related Materials