EMAIL THIS PAGE TO A FRIEND
Parasitology research
Synergistic effects of pyrantel and the febantel metabolite fenbendazole on adult Toxocara canis.
PMID 12928887
Abstract
Pyrantel embonate and febantel are both constituents of Drontal Plus and Drontal Puppy broad spectrum anthelmintics for dogs. The effects of pyrantel and the febantel metabolite fenbendazole were investigated against Toxocara canis in-vitro by studying changes in worm motility and tissue damage. Pyrantel and fenbendazole were added to worms incubated in media for 8 h at the following concentrations: pyrantel: 12.2 microg, 25 microg, or 50 microg; fenbendazole: 50 microg, 100 microg or 200 microg; mixture of pyrantel and fenbendazole: 12.2 microg p + 50 microg f, 25 microg p + 100 microg f, 50 microg p + 200 microg f. Following this 8 h incubation period, one group of the worms was immediately fixed and studied by light- and electron microscopical examination. Other groups have been observed for further 8 h periods up to 56 hours and then studied in the same way.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33981
Febantel, VETRANAL, analytical standard
C20H22N4O6S