EMAIL THIS PAGE TO A FRIEND
Journal of Neuroscience Research
Chronic nicotine administration increases NGF-like immunoreactivity in frontoparietal cerebral cortex.
PMID 12929138
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB1526P
Anti-Nerve Growth Factor-β Antibody, pain, Chemicon®, from rabbit