EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A long-range chiral relay via tertiary amide group in asymmetric catalysis: new amino acid-derived N,P-ligands for copper-catalysed conjugate addition.
PMID 12932047
Abstract
New N,P-ligands 4-6, derived from valinol and prolinol, respectively, have been developed for the asymmetric, copper(I)-catalysed conjugate addition of diethylzinc to unsaturated ketones; the tertiary amide group has been shown to effectively relay the chiral information from the ligand backbone to the active centre.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
184837
2-Amino-3-methyl-1-butanol, 97%
C5H13NO