EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Unusually high phosphodiesterolytic activity of La(III) hydroxide complexes stabilized by glycine derivatives.
PMID 12932057
Abstract
Glycine and N,N-dimethylglycine stabilize La(III) hydroxide complexes of the type La2L2(OH)4 which possess phosphodiesterolytic activity close to that observed with most active tetravalent cations like Ce(IV).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447226
Lanthanum(III) hydroxide, 99.9% trace metals basis
H3LaO3