EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Decomposition of 2-bromophenol in NaOH solution at high temperature.
PMID 12935756
Abstract
2-Bromophenol was reacted in aqueous sodium hydroxide at 200-250 degrees C. The decomposition rate was remarkably faster at 250 degrees C than at 225 or 200 degrees C, and the percentage debromination reached almost 100% in 1M NaOH at 250 degrees C for 4h. The percentage increased with NaOH concentration over the range 0.1-1M. Aliphatic compounds, such as 2,2-dimethoxypropane and 4-hydroxy-4-methyl-2-pentanone, and aromatic compounds, such as phenol and cresol, were formed as decomposition products. The formation of carboxylic acids, such as formic, acetic, and propionic acids, in the presence of oxygen was also confirmed. Under a nitrogen atmosphere, the oxidation caused by oxygen in solution was suppressed and hydrolysis became the dominant reaction in the decomposition of 2-bromophenol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130915
2-Bromophenol, 98%
C6H5BrO