EMAIL THIS PAGE TO A FRIEND
FEBS letters
Anti-herbicide single-chain antibody expression confers herbicide tolerance in transgenic plants.
PMID 12935907
Abstract
An anti-chlorpropham single-chain variable-fragment (scFv) gene was introduced into Arabidopsis in a manner to express the antibody fragment in each of four different subcellular compartments. The accumulation of scFv in transgenic plants was detected by targeting the fragment in the endoplasmic reticulum or apoplastic space, or by expressing the fragment as a glycosylphosphatidylinositol-anchored protein, while no accumulation could be detected by targeting the fragment in the cytosol. Transgenic plants accumulating the scFv gene at a high level in the endoplasmic reticulum had enhanced tolerance to chlorpropham in comparison with the non-transformants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45393
Chlorpropham, PESTANAL®, analytical standard
C10H12ClNO2