EMAIL THIS PAGE TO A FRIEND
Organic letters
Protection of the amino group of adenosine and guanosine derivatives by elaboration into a 2,5-dimethylpyrrole moiety.
PMID 12943423
Abstract
[reaction: see text] Protection of the amino group of adenine and guanine nucleosides was effected by heating the substrates in 2,5-hexanedione. The resulting 2,5-dimethylpyrrole adducts were stable toward bases but were hydrolyzed with TFA/H(2)O to regenerate the amino function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D183601
2,5-Dimethylpyrrole, 98%
C6H9N