EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Minimal methylated substrate and extended substrate range of Escherichia coli AlkB protein, a 1-methyladenine-DNA dioxygenase.
PMID 12944387
Abstract
The Escherichia coli AlkB protein, and two human homologs ABH2 and ABH3, directly demethylate 1-methyladenine and 3-methylcytosine in DNA. They couple Fe(II)-dependent oxidative demethylation of these damaged bases to decarboxylation of alpha-ketoglutarate. Here, we have determined the kinetic parameters for AlkB oxidation of 1-methyladenine in poly(dA), short oligodeoxyribonucleotides, nucleotides, and nucleoside triphosphates. Methylated poly(dA) was the preferred AlkB substrate of those tested. The oligonucleotide trimer d(Tp1meApT) and even 5'-phosphorylated 1-me-dAMP were relatively efficiently demethylated, and competed with methylated poly(dA) for AlkB activity. A polynucleotide structure was clearly not essential for AlkB to repair 1-methyladenine effectively, but a nucleotide 5' phosphate group was required. Consequently, 1-me-dAMP(5') was identified as the minimal effective AlkB substrate. The nucleoside triphosphate, 1-me-dATP, was inefficiently but actively demethylated by AlkB; a reaction with 1-me-ATP was even slower. E. coli DNA polymerase I Klenow fragment could employ 1-me-dATP as a precursor for DNA synthesis in vitro, suggesting that demethylation of alkylated deoxynucleoside triphosphates by AlkB could have biological significance. Although the human enzymes, ABH2 and ABH3, demethylated 1-methyladenine residues in poly(dA), they were inefficient with shorter substrates. Thus, ABH3 had very low activity on the trimer, d(Tp1meApT), whereas no activity was detected with ABH2. AlkB is known to repair methyl and ethyl adducts in DNA; to extend this substrate range, AlkB was shown to reduce the toxic effects of DNA damaging agents that generate hydroxyethyl, propyl, and hydroxypropyl adducts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RECOMT
Thrombin CleanCleave Kit