EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and structure-activity relationships of 1-arylmethyl-3-(1-methyl-2-amino)ethyl-5-aryl-6-methyluracils as antagonists of the human GnRH Receptor.
PMID 12951117
Abstract
A new class of small molecule GnRH antagonists, the 1-arylmethyl-3-(1-methyl-2-amino)ethyl-5-aryl-6-methyluracils, was designed and a novel stereoselective synthesis for these compounds was developed. The stereochemical integrities of key intermediates (S)-6 and (R)-6 were confirmed by a combination of X-ray crystallography and chiral HPLC determinations. SAR studies were performed, which allowed the identification of derivatives (R)-9f, (R)-9h and (R)-12 as potent hGnRH antagonists (K(i)=20 nM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D115207
6-Methyluracil, 97%
C5H6N2O2