EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Expression of CD38 in human promyelocytic leukemia HL-60 cell line during differentiation by niacin-related compounds.
PMID 12951526
Abstract
It was found that three niacin-related compounds, isonicotinic acid, nicotinamide, and nicotinamide N-oxide, induced granulocytic differentiation in HL-60 cells. We investigated the expression of CD38, which catalyzes the synthesis of cyclic ADP-ribose, a Ca2+ mobilizer, during differentiation by niacin-related compounds. It was found that CD38 was induced by isonicotinic acid, whereas nicotinamide and nicotinamide N-oxide containing an amino group did not induce it. The difference in expression of CD38 may provide some useful information for the elucidation of the mechanisms of cell differentiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3258
Nicotinamide N-oxide, ≥98%
C6H6N2O2