EMAIL THIS PAGE TO A FRIEND
Agents and actions
Effect of adjuvant arthritis on the disposition of acebutolol enantiomers in rats.
PMID 1295378
Abstract
Disease states such as arthritis may interact with the kinetics of beta-blockers. Acebutolol (AC) is a chiral beta-blocker which is available as a racemate. The beneficial properties of AC, however, is attributed mainly to the S-(+)-enantiomer. The disposition of AC enantiomers and their active, chiral metabolites, diacetolol (DC) were examined after oral administration to healthy and adjuvant-induced arthritic (AA) female Sprague-Dawley rats. Arthritis was induced by tail base injection of Mycobacterium butyricum. Swelling of hind and forepaws were apparent in 10-16 days in AA but not controls. Control and AA rats were sacrificed at 0.5, 1, 2, 4, 6 and 8 h after a 25 mg/kg oral AC dose and blood was collected (n = 6). Significant three to tenfold increases in the initial plasma concentrations (0.5-2 h) of AC were observed in AA. Enantiomers were equally affected, thus AC S:R ratio was not changed. Higher plasma concentrations of the metabolite were only significant at 2 h. The ratio of DC:AC, however, was unaffected by AA. The DC S:R ratio was significantly decreased at 0.5 and 1 h in AA. The limited protein binding of AC (10%) was neither stereoselective nor affected by AA. Reduced intrinsic clearance in AA may be responsible for these observations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000359 Acebutolol impurity B, European Pharmacopoeia (EP) Reference Standard
C16H24N2O4